3-Methyl-2-thiophenboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR4215-1G
APOSOR4215-1GEA
36.4
EUR
APOSOR4215-1G
3-Methyl-2-thiophenboronsäure
3-Methyl-2-thiopheneboronic acid
Formel:
C₅H₇BO₂S Schmelzpunkt: 192…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236049 CAS-Nummer: 177735-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...