2-Amino-5-iodpyrimidin 97%

Lieferant: Apollo Scientific
Achtung

OR42151-25G
APOSOR42151-25GEA 168 EUR
APOSOR42151-25G
2-Amino-5-iodpyrimidin 97%
2-Amino-5-iodopyrimidine
Formel: C₄H₄IN₃
Molecular Weight: 221 g/mol
Schmelzpunkt: 217…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075666
CAS-Nummer: 1445-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR