3-Methylisoquinoline-5-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Methyl-2-azanaphthalene-5-sulphonyl chloride, 5-(Chlorosulphonyl)-3-methylisoquinoline

OR42157-1G
APOSOR42157-1GEA 857 EUR
APOSOR42157-1G
3-Methylisoquinoline-5-sulphonyl chloride
3-Methylisoquinoline-5-sulphonyl chloride
Formel: C₁₀H₈ClNO₂S
Molecular Weight: 241.694 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052648
CAS-Nummer: 640290-02-4
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR