4-(Hydroxymethyl)-1-methylpyrrolidin-2-one

Lieferant: Apollo Scientific
Achtung

OR42168-1G
APOSOR42168-1GEA 119 EUR
APOSOR42168-1G
4-(Hydroxymethyl)-1-methylpyrrolidin-2-one
4-(Hydroxymethyl)-1-methylpyrrolidin-2-one
Formel: C₆H₁₁NO₂
Siedepunkt: 122…123 °C/0,08mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09055222
CAS-Nummer: 59887-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR