3-Methoxypiperidin-2-one

Lieferant: Apollo Scientific
Achtung

OR42169-1G
APOSOR42169-1GEA 0 EUR
APOSOR42169-1G
3-Methoxypiperidin-2-one
3-Methoxypiperidin-2-one
Formel: C₆H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12026381
CAS-Nummer: 25219-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR