3-Chlor-4-(dimethylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR4217-250MG
APOSOR4217-250MGEA
29
EUR
APOSOR4217-250MG
3-Chlor-4-(dimethylcarbamoyl)phenylboronsäure 98%
3-Chlor-4-(dimethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BClNO₃ Schmelzpunkt: 126…132 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363772 CAS-Nummer: 850589-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...