5-Brom-3-nitro-2-picolin 97%
Lieferant: Apollo Scientific
Synonyme:
5-Brom-2-methyl-3-nitropyridin
OR42176-25G
APOSOR42176-25GEA
65.4
EUR
APOSOR42176-25G
5-Brom-3-nitro-2-picolin 97%
5-Brom-3-nitro-2-picolin
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09031419 CAS-Nummer: 911434-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...