1,6-Naphthyridin

Lieferant: Apollo Scientific

Synonyme: Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene

OR42177-1G
APOSOR42177-1GEA 148 EUR
APOSOR42177-1G
1,6-Naphthyridin
1,6-Naphthyridine
Formel: C₈H₆N₂
Molecular Weight: 130,15 g/mol
Schmelzpunkt: 35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059750
CAS-Nummer: 253-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR