3-Chlor-4-(ethylcarbamoyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
OR4218-10G
OR4218-1G
OR4218-5G
APOSOR4218-10GEA
1050
EUR
APOSOR4218-10G
APOSOR4218-1G
APOSOR4218-5G
3-Chlor-4-(ethylcarbamoyl)phenylboronsäure 97%
3-Chlor-4-(ethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BClNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363767 CAS-Nummer: 850589-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...