Benzophenonimin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzophenoneimine
OR42185-100G
OR42185-25G
OR42185-500G
APOSOR42185-100GEA
62.5
EUR
APOSOR42185-100G
APOSOR42185-25G
APOSOR42185-500G
Benzophenonimin 97%
Benzophenonimin
Formel:
C₁₃H₁₁N Molecular Weight: 181.24 g/mol Siedepunkt: 151…153 °C/10mmHg Dichte: 1.08 g/cm³ (25 °C) Flammpunkt: 109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001760 CAS-Nummer: 1013-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...