5-Chlorchinolin
Lieferant: Apollo Scientific
|
Achtung
|
OR42188-1G
OR42188-25G
OR42188-5G
APOSOR42188-1GEA
72.5
EUR
APOSOR42188-1G
APOSOR42188-25G
APOSOR42188-5G
5-Chlorchinolin
5-Chloroquinoline
Formel:
C₉H₆ClN Schmelzpunkt: 28…32 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00239413 CAS-Nummer: 635-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...