3-(2-Hydroxyprop-2-yl)pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 2-(Pyridin-3-yl)propan-2-ol

OR42205-250MG
APOSOR42205-250MGEA 49.3 EUR
APOSOR42205-250MG
3-(2-Hydroxyprop-2-yl)pyridine 97%
3-(2-Hydroxyprop-2-yl)pyridine
Formel: C₈H₁₁NO
Molecular Weight: 137,179 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09966040
CAS-Nummer: 15031-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR