N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-threonin
Lieferant: Apollo Scientific
Synonyme:
N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid
OR42214-100G
OR42214-25G
OR42214-500G
APOSOR42214-100GEA
105
EUR
APOSOR42214-100G
APOSOR42214-25G
APOSOR42214-500G
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-threonin
N-Boc-L-threonine
Formel:
C₉H₁₇NO₅ Schmelzpunkt: 80…82 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065946 CAS-Nummer: 2592-18-9 EINECS: 219-987-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...