3-Chlor-4-(methylcarbamoyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-4-(N-methylcarbamoyl)phenylboronic acid, (3-Chloro-4-(methylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(methylcarbamoyl)benzeneboronic acid
OR4222-1G
APOSOR4222-1GEA
85.7
EUR
APOSOR4222-1G
3-Chlor-4-(methylcarbamoyl)phenylboronsäure 95%
3-Chlor-4-(methylcarbamoyl)phenylboronsäure
Formel:
C₈H₉BClNO₃ Molecular Weight: 213,43 g/mol Schmelzpunkt: 164…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363766 CAS-Nummer: 850589-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...