4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ethyl-5-(4-formylphenyl)-1,3,4-oxadiazole

OR42226-1G
APOSOR42226-1GEA 248 EUR
APOSOR42226-1G
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Formel: C₁₁H₁₀N₂O₂
Molecular Weight: 202,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19104824
CAS-Nummer: 1309875-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR