6-Chloroquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-Carboxy-6-chloroquinoline

OR42243-250MG
APOSOR42243-250MGEA 88.6 EUR
APOSOR42243-250MG
6-Chloroquinoline-3-carboxylic acid
6-Chloroquinoline-3-carboxylic acid
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,613 g/mol
Schmelzpunkt: 228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09787826
CAS-Nummer: 118791-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR