3-Amino-6-bromoquinoline dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 6-Bromoquinolin-3-amine dihydrochloride

OR42244-1G
APOSOR42244-1GEA 549 EUR
APOSOR42244-1G
3-Amino-6-bromoquinoline dihydrochloride
3-Amino-6-bromoquinoline dihydrochloride
Formel: C₉H₇BrN₂·2ClH
Molecular Weight: 295,991 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703347
CAS-Nummer: 1171078-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR