3-Amino-7-bromoquinoline dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 7-Bromoquinolin-3-amine dihydrochloride

OR42245-1G
APOSOR42245-1GEA 632 EUR
APOSOR42245-1G
3-Amino-7-bromoquinoline dihydrochloride
3-Amino-7-bromoquinoline dihydrochloride
Formel: C₉H₇BrN₂·2ClH
Molecular Weight: 295,991 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375329
CAS-Nummer: 1266226-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR