3-Amino-8-bromoquinoline dihydrochloride

Lieferant: Apollo Scientific

OR42246-1G
APOSOR42246-1GEA 632 EUR
APOSOR42246-1G
3-Amino-8-bromoquinoline dihydrochloride
3-Amino-8-bromoquinoline dihydrochloride
Formel: C₉H₇BrN₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18375330
CAS-Nummer: 1266226-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR