3-Amino-6-methoxyquinoline dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 6-Methoxyquinolin-3-amine dihydrochloride

OR42247-1G
APOSOR42247-1GEA 542 EUR
APOSOR42247-1G
3-Amino-6-methoxyquinoline dihydrochloride
3-Amino-6-methoxyquinoline dihydrochloride
Formel: C₁₀H₁₀N₂O·2ClH
Molecular Weight: 247,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07700222
CAS-Nummer: 1170974-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR