3-Amino-7-methoxyquinoline dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 7-Methoxyquinolin-3-amine dihydrochloride

OR42248-1G
APOSOR42248-1GEA 632 EUR
APOSOR42248-1G
3-Amino-7-methoxyquinoline dihydrochloride
3-Amino-7-methoxyquinoline dihydrochloride
Formel: C₁₀H₁₀N₂O·2ClH
Molecular Weight: 247,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14525835
CAS-Nummer: 1216228-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR