8-Bromoquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR42249-1G
APOSOR42249-1GEA
347
EUR
APOSOR42249-1G
8-Bromoquinoline-3-carboxylic acid
8-Bromoquinoline-3-carboxylic acid
Formel:
C₁₀H₆BrNO₂ Molecular Weight: 252,0641 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375331 CAS-Nummer: 347146-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...