Tetrakis(triphenylphosphin)palladium(0)
Lieferant: Apollo Scientific
|
Achtung
|
OR4225-1G
OR4225-25G
OR4225-5G
APOSOR4225-1GEA
27.7
EUR
APOSOR4225-1G
APOSOR4225-25G
APOSOR4225-5G
Tetrakis(triphenylphosphin)palladium(0)
Tetrakis(triphenylphosphin)palladium(0)
Formel:
C₇₂H₆₀P₄Pd Schmelzpunkt: 103…107 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010012 CAS-Nummer: 14221-01-3 EINECS: 238-086-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...