7-Methoxyquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
3-Carboxy-7-methoxyquinoline
OR42251-1G
APOSOR42251-1GEA
410
EUR
APOSOR42251-1G
7-Methoxyquinoline-3-carboxylic acid
7-Methoxyquinoline-3-carboxylic acid
Formel:
C₁₁H₉NO₃ Molecular Weight: 203,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787873 CAS-Nummer: 474659-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...