1-Bromo-4-ethoxy-2-methylbenzene 97%
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-2-methyl-4-ethoxybenzene
OR42253-5G
APOSOR42253-5GEA
122
EUR
APOSOR42253-5G
1-Bromo-4-ethoxy-2-methylbenzene 97%
1-Bromo-4-ethoxy-2-methylbenzene
Formel:
C₉H₁₁BrO Molecular Weight: 215,087 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18917736 CAS-Nummer: 68155-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...