3-Amino-4-brom-1H-pyrazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole
OR42254-5G
APOSOR42254-5GEA
21.9
EUR
APOSOR42254-5G
3-Amino-4-brom-1H-pyrazol 98%
3-Amino-4-bromo-1H-pyrazole
Formel:
C₃H₄BrN₃ Molecular Weight: 161,99 g/mol Schmelzpunkt: 135…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082728 CAS-Nummer: 16461-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...