5-Ethoxy-1H-indazole-6-boronic acid

Lieferant: Apollo Scientific
Achtung

OR42257-1G OR42257-500MG
APOSOR42257-1GEA 1860 EUR
APOSOR42257-1G APOSOR42257-500MG
5-Ethoxy-1H-indazole-6-boronic acid
5-Ethoxy-1H-indazole-6-boronic acid
Formel: C₉H₁₁BN₂O₃
Molecular Weight: 206.006 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22376778
CAS-Nummer: 2304634-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR