5-Isopropoxy-1H-indazole-6-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Isopropoxy-1H-indazol-6-yl)boronic acid, [5-(Propan-2-yloxy)-1H-indazol-6-yl]boronic acid
OR42258-1G
OR42258-500MG
APOSOR42258-1GEA
683
EUR
APOSOR42258-1G
APOSOR42258-500MG
5-Isopropoxy-1H-indazole-6-boronic acid
5-Isopropoxy-1H-indazole-6-boronic acid
Formel:
C₁₀H₁₃BN₂O₃ Molecular Weight: 220.033 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22376779 CAS-Nummer: 2304634-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...