5-Isopropoxy-1H-indazole-6-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Isopropoxy-1H-indazol-6-yl)boronic acid, [5-(Propan-2-yloxy)-1H-indazol-6-yl]boronic acid

OR42258-1G OR42258-500MG
APOSOR42258-1GEA 683 EUR
APOSOR42258-1G APOSOR42258-500MG
5-Isopropoxy-1H-indazole-6-boronic acid
5-Isopropoxy-1H-indazole-6-boronic acid
Formel: C₁₀H₁₃BN₂O₃
Molecular Weight: 220.033 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22376779
CAS-Nummer: 2304634-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR