Ethyl 5-(4-methoxyphenyl)isoxazole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR42269-1G
APOSOR42269-1GEA
36.4
EUR
APOSOR42269-1G
Ethyl 5-(4-methoxyphenyl)isoxazole-3-carboxylate
Ethyl 5-(4-methoxyphenyl)isoxazole-3-carboxylate
Formel:
C₁₃H₁₃NO₄ Molecular Weight: 247,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08446406 CAS-Nummer: 925006-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...