2-Amino-6-cyclopropylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Cyclopropylpyridin-2-amine

OR42286-1G OR42286-250MG
APOSOR42286-1GEA 427 EUR
APOSOR42286-1G APOSOR42286-250MG
2-Amino-6-cyclopropylpyridine
2-Amino-6-cyclopropylpyridine
Formel: C₈H₁₀N₂
Molecular Weight: 134.178 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12032770
CAS-Nummer: 857292-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR