2-Amino-6-cyclopropylpyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Cyclopropylpyridin-2-amine
OR42286-1G
OR42286-250MG
APOSOR42286-1GEA
427
EUR
APOSOR42286-1G
APOSOR42286-250MG
2-Amino-6-cyclopropylpyridine
2-Amino-6-cyclopropylpyridine
Formel:
C₈H₁₀N₂ Molecular Weight: 134.178 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12032770 CAS-Nummer: 857292-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...