1-Chloro-9-iodo-phenazine
Lieferant: Apollo Scientific
|
Achtung
|
OR42290-1G
OR42290-250MG
OR42290-5G
APOSOR42290-1GEA
108
EUR
APOSOR42290-1G
APOSOR42290-250MG
APOSOR42290-5G
1-Chloro-9-iodo-phenazine
1-Chloro-9-iodo-phenazine
Formel:
C₁₂H₆ClIN₂ Molecular Weight: 340.547 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32111680 CAS-Nummer: 2367002-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...