N-Benzyl-3-borono-4-methylbenzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Benzylsulphamoyl-2-methylbenzeneboronic acid

OR4234-250MG
APOSOR4234-250MGEA 143 EUR
APOSOR4234-250MG
N-Benzyl-3-borono-4-methylbenzolsulfonamid 97%
N-Benzyl-3-borono-4-methylbenzolsulfonamid
Formel: C₁₄H₁₆BNO₄S
Molecular Weight: 305,16 g/mol
Schmelzpunkt: 118…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363760
CAS-Nummer: 871329-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR