4-Methylcyclohexen-1-ylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Methylcyclohex-1-en-1-yl)boronic acid, 4-Methyl-1-cyclohexen-1-ylboronic acid
OR4241-250MG
APOSOR4241-250MGEA
52.2
EUR
APOSOR4241-250MG
4-Methylcyclohexen-1-ylboronsäure 98%
4-Methylcyclohexene-1-ylboronic acid
Formel:
C₇H₁₃BO₂ Molecular Weight: 139,99 g/mol Schmelzpunkt: 162…166 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06659857 CAS-Nummer: 850567-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...