[(4-Methylphenyl)thio]acetic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: [(4-Methylphenyl)sulphanyl]acetic acid, 2-(p-Tolylthio)acetic acid, 4-[(Carboxymethyl)sulphanyl]toluene

OR4248-5G
APOSOR4248-5GEA 31.9 EUR
APOSOR4248-5G
[(4-Methylphenyl)thio]acetic acid 98%
[(4-Methylphenyl)thio]acetic acid
Formel: C₉H₁₀O₂S
Molecular Weight: 182,24 g/mol
Schmelzpunkt: 79…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00034675
CAS-Nummer: 3996-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR