Benzaldehydoxim 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: alpha-benzaldoxime, Benzaldoxime

OR4290-100G
APOSOR4290-100GEA 94.4 EUR
APOSOR4290-100G
Benzaldehydoxim 90%
Benzaldehydoxim
Formel: C₇H₇NO
Molecular Weight: 121.14 g/mol
Siedepunkt: 118…120 °C/10mmHg
Dichte: 1.11 g/cm³ (25 °C)
Flammpunkt: 108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002119
CAS-Nummer: 932-90-1
EINECS: 213-261-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR