4-Chlorphenylhydraziniumchlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlorphenylhydrazinhydrochlorid, 4-Chlorphenylhydrazin Hydrochlorid, 1-(4-Chlorphenyl)hydrazin Hydrochlorid

OR4295-500G
APOSOR4295-500GEA 72.5 EUR
APOSOR4295-500G
4-Chlorphenylhydraziniumchlorid 97%
4-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
Molecular Weight: 179.05 g/mol
Schmelzpunkt: 216 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012943
CAS-Nummer: 1073-70-7
EINECS: 214-030-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR