5-Amino-2-brombenzoesäure

Lieferant: Apollo Scientific
Gefahr

OR4307-1G
APOSOR4307-1GEA 21.9 EUR
APOSOR4307-1G
5-Amino-2-brombenzoesäure
5-Amino-2-brombenzoesäure
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 171…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368968
CAS-Nummer: 2840-02-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR