2-Bromo-4-methylaniline hydrochloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Amino-3-bromotoluene hydrochloride
OR4327-5G
APOSOR4327-5GEA
69.6
EUR
APOSOR4327-5G
2-Bromo-4-methylaniline hydrochloride
2-Bromo-4-methylaniline hydrochloride
Formel:
C₇H₈BrN·ClH Molecular Weight: 222,51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142869 CAS-Nummer: 13194-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...