1-Bromo-3,5,7-trimethyladamantane
Lieferant: Apollo Scientific
|
Achtung
|
OR4338-1G
APOSOR4338-1GEA
748
EUR
APOSOR4338-1G
1-Bromo-3,5,7-trimethyladamantane
1-Bromo-3,5,7-trimethyladamantane
Formel:
C₁₃H₂₁Br Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154002 CAS-Nummer: 53398-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...