N-Boc-trans-4-Hydroxy-L-prolinmethylester

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

OR43520-500G
APOSOR43520-500GEA 98.6 EUR
APOSOR43520-500G
N-Boc-trans-4-Hydroxy-L-prolinmethylester
N-Boc-trans-4-Hydroxy-L-prolinmethylester
Formel: C₁₁H₁₉NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00076981
CAS-Nummer: 74844-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR