2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid 97%

Lieferant: Apollo Scientific

OR43534-250MG
APOSOR43534-250MGEA 197 EUR
APOSOR43534-250MG
2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid 97%
2-(Ethoxycarbonyl)-1H-indole-3-carboxylic acid
Formel: C₁₂H₁₁NO₄
Schmelzpunkt: 262…263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18803524
CAS-Nummer: 441800-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR