Methyl 2-bromo-5-methylthiophene-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-3-(methoxycarbonyl)-5-methylthiophene
OR43540-250MG
APOSOR43540-250MGEA
49.3
EUR
APOSOR43540-250MG
Methyl 2-bromo-5-methylthiophene-3-carboxylate
Methyl 2-bromo-5-methylthiophene-3-carboxylate
Formel:
C₇H₇BrO₂S Molecular Weight: 235,098 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09702018 CAS-Nummer: 944709-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...