Ethyl-2-(5-morpholino-2H-1,2,3,4-tetraazol-2-yl)acetate

Lieferant: Apollo Scientific
Achtung

OR4355-100MG
APOSOR4355-100MGEA 45.1 EUR
APOSOR4355-100MG
Ethyl-2-(5-morpholino-2H-1,2,3,4-tetraazol-2-yl)acetate
Ethyl-2-(5-morpholino-2H-1,2,3,4-tetraazol-2-yl)acetate
Formel: C₉H₁₅N₅O₃
Molecular Weight: 241.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052626
CAS-Nummer: 175205-05-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR