2-(5-Amino-1H-indazol-1-yl)ethanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-1-(2-hydroxyethyl)-1H-indazole

OR43551-1G OR43551-250MG
APOSOR43551-1GEA 435 EUR
APOSOR43551-1G APOSOR43551-250MG
2-(5-Amino-1H-indazol-1-yl)ethanol 97%
2-(5-Amino-1H-indazol-1-yl)ethanol
Formel: C₉H₁₁N₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 885270-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR