6-Brom-3-chlor-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR43556-250MG
APOSOR43556-250MGEA 29 EUR
APOSOR43556-250MG
6-Brom-3-chlor-1H-indazol
6-Bromo-3-chloro-1H-indazole
Formel: C₇H₄BrClN₂
Molecular Weight: 231,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738283
CAS-Nummer: 885271-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR