1-Boc-4-bromo-1H-indazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-Bromo-1H-indazole, 4-Bromo-1H-indazole, N1-BOC protected
OR43563-1G
APOSOR43563-1GEA
55.1
EUR
APOSOR43563-1G
1-Boc-4-bromo-1H-indazole 95%
1-Boc-4-bromo-1H-indazole
Formel:
C₁₂H₁₃BrN₂O₂ Molecular Weight: 297.1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110271 CAS-Nummer: 926922-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...