7-Bromoindazole-1-carboxylic acid tert-butyl ester 98%
Lieferant: Apollo Scientific
Synonyme:
7-Bromo-1H-indazole, N1-BOC protected, tert-Butyl 7-bromo-1H-indazole-1-carboxylate
OR43565-250MG
APOSOR43565-250MGEA
85.7
EUR
APOSOR43565-250MG
7-Bromoindazole-1-carboxylic acid tert-butyl ester 98%
7-Bromoindazole-1-carboxylic acid tert-butyl ester
Formel:
C₁₂H₁₃BrN₂O₂ Molecular Weight: 297,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505878 CAS-Nummer: 1092352-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...