5-Brom-2-methyl-2H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-5-bromo-2H-indazole

OR43568-250MG
APOSOR43568-250MGEA 21.9 EUR
APOSOR43568-250MG
5-Brom-2-methyl-2H-indazol
5-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,0600 g/mol
Schmelzpunkt: 97…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09870047
CAS-Nummer: 465529-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR